Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002666.1 | NP_001002666.2 | 11 - 12 | 1458 | 82 | |
GenBank mRNA | BC076553.1 | - | 11 - 12 | 1458 | 82 | |
BC163973.1 | - | 11 - 12 | 1372 | 82 | ||
HM138690.1 | - | 11 - 12 | 1344 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |