Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pbrm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003535.2 | NP_001003535.2 | 6 - 7 | 780 | 90 | |
XM_005168762.4 | XP_005168819.1 | 6 - 7 | 780 | 90 | ||
GenBank mRNA | BC078299.1 | - | 6 - 7 | 784 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |