Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cibar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003604.2 | NP_001003604.1 | 2 - 3 | 360 | 143 | |
XM_021468050.1 | XP_021323725.1 | 2 - 3 | 399 | 143 | ||
GenBank mRNA | BC078261.1 | - | 2 - 3 | 364 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |