Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004119.1 | NP_001004119.1 | 2 - 3 | 239 | 102 | |
XM_021470381.1 | XP_021326056.1 | 2 - 3 | 265 | 102 | ||
GenBank mRNA | BC080211.1 | - | 2 - 3 | 239 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |