Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ap1s3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004635.1 | NP_001004635.1 | 4 - 5 | 523 | 81 | |
GenBank mRNA | BC081385.1 | - | 4 - 5 | 523 | 81 | |
BC150444.1 | - | 4 - 5 | 492 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |