Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rrh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004654.1 | NP_001004654.1 | 6 - 7 | 957 | 82 | |
GenBank mRNA | BC081524.1 | - | 6 - 7 | 957 | 82 | |
BC164135.1 | - | 6 - 7 | 932 | 82 | ||
KT008425.1 | - | 6 - 7 | 898 | 82 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |