Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trpa1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007065.1 | NP_001007066.1 | 27 - 28 | 3164 | 68 | |
XM_017358267.2 | XP_017213756.1 | 26 - 27 | 3159 | 68 | ||
GenBank mRNA | AY677196.1 | - | 27 - 28 | 3164 | 68 | |
EU826643.1 | - | 26 - 27 | 3133 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Danio rerio Probe spans exons |