Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014345.2 | NP_001014367.2 | 5 - 6 | 700 | 116 | |
GenBank mRNA | AY880251.1 | - | 5 - 6 | 700 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |