Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ing4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020468.2 | NP_001018304.1 | 7 - 8 | 744 | 65 | |
GenBank mRNA | BC095338.1 | - | 7 - 8 | 740 | 65 | |
BC165444.1 | - | 7 - 8 | 750 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |