Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020526.2 | NP_001018362.1 | 4 - 5 | 554 | 72 | |
GenBank mRNA | BC095041.1 | - | 4 - 5 | 554 | 72 | |
BC164300.1 | - | 4 - 5 | 471 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |