Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stxbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025182.1 | NP_001020353.1 | 11 - 12 | 992 | 69 | |
XM_005161113.4 | XP_005161170.1 | 11 - 12 | 1067 | 69 | ||
GenBank mRNA | BC097221.1 | - | 11 - 12 | 992 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |