Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nitr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025496.1 | NP_001020667.1 | 1 - 2 | 42 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |