Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nitr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025496.1 | NP_001020667.1 | 4 - 5 | 885 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Danio rerio Probe spans exons |