Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025538.2 | NP_001020709.1 | 6 - 7 | 773 | 55 | |
GenBank mRNA | BC097114.1 | - | 6 - 7 | 770 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |