Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2ad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025554.2 | NP_001020725.1 | 1 - 2 | 264 | 117 | |
GenBank mRNA | AY500819.1 | - | 1 - 2 | 184 | 117 | |
BC085624.1 | - | 1 - 2 | 237 | 117 | ||
BC124073.1 | - | 1 - 2 | 231 | 117 | ||
BC134877.1 | - | 1 - 2 | 264 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |