Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2ad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025554.2 | NP_001020725.1 | 3 - 4 | 579 | 94 | |
GenBank mRNA | AY500819.1 | - | 3 - 4 | 499 | 94 | |
BC085624.1 | - | 3 - 4 | 552 | 94 | ||
BC124073.1 | - | 3 - 4 | 546 | 94 | ||
BC134877.1 | - | 3 - 4 | 579 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |