Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2ad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025554.2 | NP_001020725.1 | 7 - 8 | 1246 | 80 | |
GenBank mRNA | BC085624.1 | - | 6 - 7 | 1393 | 80 | |
BC124073.1 | - | 6 - 7 | 1387 | 80 | ||
BC134877.1 | - | 7 - 8 | 1246 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |