Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rdh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030101.1 | NP_001025272.1 | 3 - 4 | 495 | 70 | |
XM_005173961.4 | XP_005174018.1 | 3 - 4 | 550 | 70 | ||
XM_009296105.3 | XP_009294380.1 | 3 - 4 | 556 | 70 | ||
GenBank mRNA | BC097151.1 | - | 3 - 4 | 495 | 70 | |
DQ000308.1 | - | 2 - 3 | 338 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |