Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc35e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030179.1 | NP_001025350.1 | 2 - 3 | 716 | 69 | |
GenBank mRNA | BC116467.1 | - | 2 - 3 | 677 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |