Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc4a4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034984.1 | NP_001030156.1 | 3 - 4 | 271 | 76 | |
XM_005165244.4 | XP_005165301.1 | 3 - 4 | 548 | 76 | ||
XM_009301742.3 | XP_009300017.1 | 2 - 3 | 479 | 76 | ||
XM_009301743.3 | XP_009300018.1 | 2 - 3 | 478 | 76 | ||
XM_009301747.3 | XP_009300022.1 | 2 - 3 | 478 | 76 | ||
XM_021476291.1 | XP_021331966.1 | 4 - 5 | 468 | 76 | ||
XM_021476293.1 | XP_021331968.1 | 4 - 5 | 441 | 76 | ||
XM_021476294.1 | XP_021331969.1 | 4 - 5 | 410 | 76 | ||
GenBank mRNA | AY727858.1 | - | 3 - 4 | 271 | 76 | |
BC162559.1 | - | 3 - 4 | 330 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |