Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc4a4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034984.1 | NP_001030156.1 | 7 - 8 | 824 | 74 | |
XM_005165244.4 | XP_005165301.1 | 7 - 8 | 1074 | 74 | ||
XM_009301742.3 | XP_009300017.1 | 7 - 8 | 1128 | 74 | ||
XM_009301743.3 | XP_009300018.1 | 6 - 7 | 1031 | 74 | ||
XM_009301747.3 | XP_009300022.1 | 6 - 7 | 1004 | 74 | ||
XM_021476291.1 | XP_021331966.1 | 9 - 10 | 1117 | 74 | ||
XM_021476293.1 | XP_021331968.1 | 9 - 10 | 1090 | 74 | ||
XM_021476294.1 | XP_021331969.1 | 9 - 10 | 1059 | 74 | ||
GenBank mRNA | AY727858.1 | - | 7 - 8 | 824 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |