Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zte38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039809.1 | NP_001034898.1 | 8 - 9 | 724 | 78 | |
GenBank mRNA | BC114280.1 | - | 8 - 9 | 724 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |