Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtmr7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040316.2 | NP_001035406.1 | 13 - 14 | 1716 | 134 | |
GenBank mRNA | BC115252.1 | - | 13 - 14 | 1717 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |