Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnfsf10l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042713.1 | NP_001036178.1 | 4 - 5 | 411 | 81 | |
GenBank mRNA | BC162834.1 | - | 4 - 5 | 493 | 81 | |
BC162841.1 | - | 4 - 5 | 493 | 81 | ||
DQ812113.1 | - | 4 - 5 | 411 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |