Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhoj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044801.2 | NP_001038266.1 | 3 - 4 | 436 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |