Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klf11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044941.2 | NP_001038406.1 | 3 - 4 | 1341 | 115 | |
XM_021468456.1 | XP_021324131.1 | 3 - 4 | 1341 | 115 | ||
GenBank mRNA | BC122208.1 | - | 3 - 4 | 1343 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |