Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snap47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045437.1 | NP_001038902.1 | 4 - 5 | 1309 | 116 | |
XM_021481304.1 | XP_021336979.1 | 4 - 5 | 1469 | 116 | ||
GenBank mRNA | BC122195.1 | - | 4 - 5 | 1309 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |