Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: isoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079953.2 | NP_001073422.2 | 3 - 4 | 278 | 75 | |
GenBank mRNA | BC125848.1 | - | 3 - 4 | 278 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |