Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scn3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080802.1 | NP_001074271.1 | 3 - 4 | 574 | 103 | |
XM_009291661.2 | XP_009289936.1 | 3 - 4 | 586 | 103 | ||
XM_009291662.2 | XP_009289937.1 | 3 - 4 | 588 | 103 | ||
GenBank mRNA | BC163285.1 | - | 3 - 4 | 471 | 103 | |
BC163295.1 | - | 3 - 4 | 471 | 103 | ||
DQ489730.1 | - | 3 - 4 | 574 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |