Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082807.1 | NP_001076276.1 | 2 - 3 | 566 | 77 | |
GenBank mRNA | BC134198.1 | - | 2 - 3 | 566 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |