Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mmab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082809.1 | NP_001076278.1 | 7 - 8 | 621 | 75 | |
GenBank mRNA | BC133925.1 | - | 7 - 8 | 574 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |