Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nog5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082988.2 | NP_001076457.1 | 1 | 356 | 75 | |
GenBank mRNA | AY779060.1 | - | 1 | 356 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |