Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083812.2 | NP_001077281.2 | 4 - 5 | 519 | 70 | |
XM_021468754.1 | XP_021324429.1 | 4 - 5 | 491 | 70 | ||
GenBank mRNA | BC085600.1 | - | 4 - 5 | 496 | 70 | |
BC108016.1 | - | 4 - 5 | 534 | 70 | ||
BC116571.1 | - | 4 - 5 | 524 | 70 | ||
BC124149.1 | - | 4 - 5 | 520 | 70 | ||
BC135070.1 | - | 4 - 5 | 493 | 70 | ||
BM096368.1 | - | 4 - 5 | 522 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |