Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089395.2 | NP_001082864.2 | 1 - 2 | 202 | 65 | |
GenBank mRNA | BC116599.1 | - | 1 - 2 | 202 | 65 | |
BC134867.1 | - | 1 - 2 | 202 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |