Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089395.2 | NP_001082864.2 | 4 - 5 | 583 | 73 | |
GenBank mRNA | BC134867.1 | - | 4 - 5 | 583 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |