Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089395.2 | NP_001082864.2 | 5 - 6 | 791 | 119 | |
GenBank mRNA | BC134867.1 | - | 5 - 6 | 791 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |