Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhobtb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099974.2 | NP_001093444.1 | 8 - 9 | 2601 | 64 | |
NM_001365558.1 | NP_001352487.1 | 8 - 9 | 2247 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |