Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhobtb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099974.2 | NP_001093444.1 | 10 - 11 | 2788 | 81 | |
NM_001365558.1 | NP_001352487.1 | 10 - 11 | 2434 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Danio rerio Probe spans exons |