Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104649.2 | NP_001098119.2 | 3 - 4 | 427 | 106 | |
XM_021471707.1 | XP_021327382.1 | 2 - 3 | 400 | 106 | ||
XM_021471716.1 | XP_021327391.1 | 3 - 4 | 489 | 106 | ||
GenBank mRNA | EU124003.1 | - | 3 - 4 | 441 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |