Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: isl2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130964.2 | NP_571039.1 | 4 - 5 | 883 | 95 | |
XM_021477390.1 | XP_021333065.1 | 3 - 4 | 675 | 95 | ||
GenBank mRNA | BC067333.1 | - | 4 - 5 | 862 | 95 | |
BC095011.1 | - | 4 - 5 | 883 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |