Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: notch1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131441.1 | NP_571516.1 | 19 - 20 | 3478 | 55 | |
GenBank mRNA | X69088.1 | - | 3478 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons |