Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eomesa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131679.3 | NP_571754.3 | 2 - 3 | 1000 | 62 | |
XM_005159319.4 | XP_005159376.1 | 3 - 4 | 62 | |||
XM_005159321.4 | XP_005159378.1 | 3 - 4 | 62 | |||
GenBank mRNA | AF287007.2 | - | 3 - 4 | 62 | ||
AF329830.1 | - | 2 - 3 | 962 | 62 | ||
BC067719.1 | - | 2 - 3 | 1000 | 62 | ||
BC154205.1 | - | 3 - 4 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |