Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eomesa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131679.3 | NP_571754.3 | 3 - 4 | 1126 | 79 | |
XM_005159319.4 | XP_005159376.1 | 4 - 5 | 1574 | 79 | ||
XM_005159321.4 | XP_005159378.1 | 4 - 5 | 1460 | 79 | ||
GenBank mRNA | AF287007.2 | - | 4 - 5 | 1030 | 79 | |
AF329830.1 | - | 3 - 4 | 1088 | 79 | ||
BC067719.1 | - | 3 - 4 | 1126 | 79 | ||
BC154205.1 | - | 4 - 5 | 1349 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |