Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131714.2 | NP_571789.2 | 5 - 6 | 565 | 74 | |
XM_005173133.2 | XP_005173190.1 | 3 - 4 | 522 | 74 | ||
GenBank mRNA | AB032415.1 | - | 5 - 6 | 618 | 74 | |
BC045280.1 | - | 5 - 6 | 565 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |