Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131879.2 | NP_571954.1 | 4 - 5 | 1143 | 64 | |
GenBank mRNA | AB078927.1 | - | 4 - 5 | 1143 | 64 | |
AF057713.1 | - | 1 - 2 | 98 | 64 | ||
AF210727.2 | - | 4 - 5 | 1118 | 64 | ||
AY398333.1 | - | 4 - 5 | 1117 | 64 | ||
BC094977.1 | - | 4 - 5 | 1143 | 64 | ||
BC165188.1 | - | 3 - 4 | 1088 | 64 | ||
CF997194.1 | - | 3 - 4 | 277 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |