Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfpia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309803.1 | NP_001296732.1 | 4 - 5 | 658 | 72 | |
NM_182872.2 | NP_878292.2 | 4 - 5 | 655 | 72 | ||
GenBank mRNA | AF515274.1 | - | 4 - 5 | 519 | 72 | |
BC133127.1 | - | 4 - 5 | 572 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |