Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdcd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198978.1 | NP_945329.1 | 9 - 10 | 1261 | 70 | |
XM_005161791.4 | XP_005161848.1 | 10 - 11 | 1344 | 70 | ||
GenBank mRNA | BC045513.1 | - | 9 - 10 | 1261 | 70 | |
BC065341.1 | - | 10 - 11 | 1286 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |