Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tsc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200052.1 | NP_956346.1 | 9 - 10 | 1165 | 76 | |
XM_005171795.4 | XP_005171852.1 | 9 - 10 | 76 | |||
XM_005171796.4 | XP_005171853.1 | 9 - 10 | 76 | |||
XM_005171797.4 | XP_005171854.1 | 9 - 10 | 76 | |||
XM_021476019.1 | XP_021331694.1 | 1 - 2 | 76 | |||
GenBank mRNA | BC055190.1 | - | 9 - 10 | 1165 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |