Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smfn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200744.2 | NP_957038.2 | 6 - 7 | 763 | 74 | |
GenBank mRNA | BC059540.1 | - | 6 - 7 | 647 | 74 | |
EH463590.1 | - | 1 - 2 | 486 | 74 | ||
EH469615.1 | - | 1 - 2 | 471 | 74 | ||
EH580645.1 | - | 6 - 7 | 797 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |