Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: surf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201127.1 | NP_957421.1 | 1 - 2 | 173 | 76 | |
GenBank mRNA | BC056295.1 | - | 1 - 2 | 173 | 76 | |
BC066509.1 | - | 1 - 2 | 159 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |